accessibility menu, dialog, popup

UserName

CH3H3CCH3OOO

CAS RN 5333-40-4

IUPAC Name: 2-[5-methyl-2-(propan-2-yl)phenoxy]acetate
Synonyms: 2-(2-isopropyl-5-methylphenoxy)acetate
Molecular Weight (g/mol): 207.25
Molecular Formula: C12H15O3
InChi Key: MURZAHIYMVFXCF-UHFFFAOYSA-M
SMILES: CC(C)C1=CC=C(C)C=C1OCC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results