missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Isopropenyltoluene (stabilized with BHT) 98.0+%, TCI America™
Supplier: TCI America I02985ML
Specifications
| 2-Isopropenyltoluene (stabilized with BHT) | |
| Colorless | |
| C10H12 | |
| 5 mL | |
| OGMSGZZPTQNTIK-UHFFFAOYSA-N | |
| 1-methyl-2-prop-1-en-2-ylbenzene | |
| 81886 | |
| ≥98.0% (GC) |
| 7399-49-7 | |
| 173°C | |
| MFCD00049092 | |
| 3295 | |
| CC1=CC=CC=C1C(=C)C | |
| 132.206 | |
| 132.21 | |
| Liquid |
Chemical Identifiers
| 7399-49-7 | |
| 132.206 | |
| OGMSGZZPTQNTIK-UHFFFAOYSA-N | |
| 1-methyl-2-prop-1-en-2-ylbenzene |
| C10H12 | |
| MFCD00049092 | |
| 81886 | |
| CC1=CC=CC=C1C(=C)C |
Safety and Handling
EINECSNumber : (3)-0010
TSCA : No