accessibility menu, dialog, popup

UserName

CAS RN 7399-49-7

H3CCH2H3C

CAS RN 7399-49-7

IUPAC Name: 1-methyl-2-(prop-1-en-2-yl)benzene
Synonyms: 1-methyl-2-ethenyl benzene
Molecular Weight (g/mol): 132.21
Molecular Formula: C10H12
InChi Key: OGMSGZZPTQNTIK-UHFFFAOYSA-N
SMILES: CC(=C)C1=CC=CC=C1C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results