missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropenyltoluene (stabilized with BHT) 98.0+%, TCI America™

Catalog No. i02981ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mL
5 mL
Catalog No. Quantity
I02981ML 1 mL
I02985ML 5 mL
2 options

Catalog No. I02981ML

Supplier: TCI America I02981ML

Only null left

Chemical Identifiers

CAS 7399-49-7
Molecular Formula C10H12
Molecular Weight (g/mol) 132.206
MDL Number MFCD00049092
InChI Key OGMSGZZPTQNTIK-UHFFFAOYSA-N
PubChem CID 81886
IUPAC Name 1-methyl-2-prop-1-en-2-ylbenzene
SMILES CC1=CC=CC=C1C(=C)C

Specifications

Color Colorless
Boiling Point 173°C
Quantity 1 mL
UN Number 3295
Formula Weight 132.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Isopropenyltoluene (stabilized with BHT)
EINECSNumber (3)-0010
TSCA No