missing translation for 'onlineSavingsMsg'
Learn More

2-Iodosobenzoic acid, 97%

CAS: 304-91-6 | C7H5IO3 | 264.018 g/mol

Supplier:  Thermo Scientific Chemicals L1560606

 View more versions of this product

Catalog No. AAL1560606


Only null left
Explore available promotions

Description

Description

1-Organosulfonyloxy-1, 2-benziodoxol-3-(1H)-ones (3, 4, 5) can be prepared in one step from 2-iodosobenzoic acid and the corresponding sulfonic acids. 2-carboxydiphenyliodonium salts was prepared by the acid-catalyzed condensation of 2-iodosobenzoic acid with benzene, mesitylene, and cyclohexylbenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Organosulfonyloxy-1, 2-benziodoxol-3-(1H)-ones (3, 4, 5) can be prepared in one step from 2-iodosobenzoic acid and the corresponding sulfonic acids. 2-carboxydiphenyliodonium salts was prepared by the acid-catalyzed condensation of 2-iodosobenzoic acid with benzene, mesitylene, and cyclohexylbenzene.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Specifications

Specifications

2-Iodosobenzoic acid
∽230°C (decomposition)
MFCD00002401
1939973
2-iodosobenzoic acid, o-iodosobenzoic acid, o-iodosylbenzoic acid, o-iodosobenzoate, benzoic acid, 2-iodosyl, benzoic acid, o-iodoso, 2-iodosyl benzoate, benzoic acid, iodosyl, ortho-iodosylbenzoic acid, 3h-1,2-benziodoxol-1-ium, 3-oxo-, hydroxide
IFPHDUVGLXEIOQ-UHFFFAOYSA-N
2-iodosylbenzoic acid
67537
264.02
304-91-6
C7H5IO3
5 g
Light sensitive
Insoluble in water.
C1=CC=C(C(=C1)C(=O)O)I=O
264.018
CHEBI:52698
97%

Chemical Identifiers

304-91-6
264.018
IFPHDUVGLXEIOQ-UHFFFAOYSA-N
67537
2-iodosylbenzoic acid
C7H5IO3
MFCD00002401
2-iodosobenzoic acid, o-iodosobenzoic acid, o-iodosylbenzoic acid, o-iodosobenzoate, benzoic acid, 2-iodosyl, benzoic acid, o-iodoso, 2-iodosyl benzoate, benzoic acid, iodosyl, ortho-iodosylbenzoic acid, 3h-1,2-benziodoxol-1-ium, 3-oxo-, hydroxide
CHEBI:52698
C1=CC=C(C(=C1)C(=O)O)I=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 206-159-4

RTECSNumber : DH3056550

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only