missing translation for 'onlineSavingsMsg'
Learn More

2-Iodosobenzoic acid, 97%

Catalog No. p-7046659
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1560603 1 g
AAL1560606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1560603 Supplier Thermo Scientific Chemicals Supplier No. L1560603
Only null left

CAS: 304-91-6 | C7H5IO3 | 264.018 g/mol

1-Organosulfonyloxy-1, 2-benziodoxol-3-(1H)-ones (3, 4, 5) can be prepared in one step from 2-iodosobenzoic acid and the corresponding sulfonic acids. 2-carboxydiphenyliodonium salts was prepared by the acid-catalyzed condensation of 2-iodosobenzoic acid with benzene, mesitylene, and cyclohexylbenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Organosulfonyloxy-1, 2-benziodoxol-3-(1H)-ones (3, 4, 5) can be prepared in one step from 2-iodosobenzoic acid and the corresponding sulfonic acids. 2-carboxydiphenyliodonium salts was prepared by the acid-catalyzed condensation of 2-iodosobenzoic acid with benzene, mesitylene, and cyclohexylbenzene.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.

Chemical Identifiers

CAS 304-91-6
Molecular Formula C7H5IO3
Molecular Weight (g/mol) 264.018
MDL Number MFCD00002401
InChI Key IFPHDUVGLXEIOQ-UHFFFAOYSA-N
Synonym 2-iodosobenzoic acid, o-iodosobenzoic acid, o-iodosylbenzoic acid, o-iodosobenzoate, benzoic acid, 2-iodosyl, benzoic acid, o-iodoso, 2-iodosyl benzoate, benzoic acid, iodosyl, ortho-iodosylbenzoic acid, 3h-1,2-benziodoxol-1-ium, 3-oxo-, hydroxide
PubChem CID 67537
ChEBI CHEBI:52698
IUPAC Name 2-iodosylbenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)I=O

Specifications

Melting Point ∽230°C (decomposition)
Quantity 1 g
Beilstein 1939973
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 264.02
Percent Purity 97%
Chemical Name or Material 2-Iodosobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-159-4
RTECSNumber DH3056550
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only