Learn More
2-Iodosobenzoic acid, 97%
CAS: 304-91-6 | C7H5IO3 | 264.018 g/mol
Supplier: Thermo Scientific Chemicals L1560603
Description
1-Organosulfonyloxy-1, 2-benziodoxol-3-(1H)-ones (3, 4, 5) can be prepared in one step from 2-iodosobenzoic acid and the corresponding sulfonic acids. 2-carboxydiphenyliodonium salts was prepared by the acid-catalyzed condensation of 2-iodosobenzoic acid with benzene, mesitylene, and cyclohexylbenzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Organosulfonyloxy-1, 2-benziodoxol-3-(1H)-ones (3, 4, 5) can be prepared in one step from 2-iodosobenzoic acid and the corresponding sulfonic acids. 2-carboxydiphenyliodonium salts was prepared by the acid-catalyzed condensation of 2-iodosobenzoic acid with benzene, mesitylene, and cyclohexylbenzene.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Specifications
| 2-Iodosobenzoic acid | |
| ∽230°C (decomposition) | |
| MFCD00002401 | |
| 1939973 | |
| 2-iodosobenzoic acid, o-iodosobenzoic acid, o-iodosylbenzoic acid, o-iodosobenzoate, benzoic acid, 2-iodosyl, benzoic acid, o-iodoso, 2-iodosyl benzoate, benzoic acid, iodosyl, ortho-iodosylbenzoic acid, 3h-1,2-benziodoxol-1-ium, 3-oxo-, hydroxide | |
| IFPHDUVGLXEIOQ-UHFFFAOYSA-N | |
| 2-iodosylbenzoic acid | |
| 67537 | |
| 264.02 |
| 304-91-6 | |
| C7H5IO3 | |
| 1 g | |
| Light sensitive | |
| Insoluble in water. | |
| C1=CC=C(C(=C1)C(=O)O)I=O | |
| 264.018 | |
| CHEBI:52698 | |
| 97% |
Chemical Identifiers
| 304-91-6 | |
| 264.018 | |
| IFPHDUVGLXEIOQ-UHFFFAOYSA-N | |
| 67537 | |
| 2-iodosylbenzoic acid |
| C7H5IO3 | |
| MFCD00002401 | |
| 2-iodosobenzoic acid, o-iodosobenzoic acid, o-iodosylbenzoic acid, o-iodosobenzoate, benzoic acid, 2-iodosyl, benzoic acid, o-iodoso, 2-iodosyl benzoate, benzoic acid, iodosyl, ortho-iodosylbenzoic acid, 3h-1,2-benziodoxol-1-ium, 3-oxo-, hydroxide | |
| CHEBI:52698 | |
| C1=CC=C(C(=C1)C(=O)O)I=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 206-159-4
RTECSNumber : DH3056550
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only