missing translation for 'onlineSavingsMsg'
Learn More

2-Iodophenylacetonitrile, 96%

Catalog No. p-7046009
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1276703 1 g
AAL1276706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1276703 Supplier Thermo Scientific Chemicals Supplier No. L1276703

CAS: 40400-15-5 | C8H6IN | 243.047 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40400-15-5
Molecular Formula C8H6IN
Molecular Weight (g/mol) 243.047
MDL Number MFCD00040888
InChI Key FPSGTRJUQLYLHE-UHFFFAOYSA-N
Synonym 2-iodophenylacetonitrile, 2-2-iodophenyl acetonitrile, 2-iodobenzyl cyanide, o-iodophenylacetonitrile, benzeneacetonitrile, 2-iodo, o-iodobenzyl cyanide, acmc-20a7xu, 2-iodo-phenylacetonitrile, 2-iodo-phenyl-acetonitrile, 2-2-iodophenyl ethanenitrile
PubChem CID 4402282
IUPAC Name 2-(2-iodophenyl)acetonitrile
SMILES C1=CC=C(C(=C1)CC#N)I

Specifications

Density 1.75
Boiling Point 139°C to 140°C (13 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.617
Quantity 1 g
UN Number UN3276
Beilstein 2083106
Sensitivity Light sensitive
Formula Weight 243.05
Percent Purity 96%
Chemical Name or Material 2-Iodophenylacetonitrile
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only