CAS RN 40400-15-5
IUPAC Name:
2-(2-iodophenyl)acetonitrile
Synonyms:
2-(2-iodophenyl)acetonitrile
Molecular Weight (g/mol):
243.05
Molecular Formula:
C8H6IN
InChi Key:
FPSGTRJUQLYLHE-UHFFFAOYSA-N
SMILES:
IC1=CC=CC=C1CC#N
1
–
1
of
1
results
1
Promotions Available
2-Iodophenylacetonitrile, 96%
2-Iodophenylacetonitrile, 96%
CAS: 40400-15-5 Molecular Formula: C8H6IN Molecular Weight (g/mol): 243.047 MDL Number: MFCD00040888 InChI Key: FPSGTRJUQLYLHE-UHFFFAOYSA-N Synonym: 2-iodophenylacetonitrile,2-2-iodophenyl acetonitrile,2-iodobenzyl cyanide,o-iodophenylacetonitrile,benzeneacetonitrile, 2-iodo,o-iodobenzyl cyanide,acmc-20a7xu,2-iodo-phenylacetonitrile,2-iodo-phenyl-acetonitrile,2-2-iodophenyl ethanenitrile PubChem CID: 4402282 IUPAC Name: 2-(2-iodophenyl)acetonitrile SMILES: C1=CC=C(C(=C1)CC#N)I