Learn More
2-Iodophenol, 98%
CAS: 533-58-4 | C6H5IO | 220.01 g/mol
Supplier: Thermo Scientific Chemicals A1359906
Description
2-Iodophenol is used in the synthesis of aryl 2-benzofuranyl and aryl 2-indolyl carbinols of high enantiomeric purity and 3,3-disubstituted-2,3-dihydrobenzofurans. Coupling with phenylacetylene in the presence of a Pd complex and CuI gives 2-phenylbenzofuran in good yield.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Iodophenol is used in the synthesis of aryl 2-benzofuranyl and aryl 2-indolyl carbinols of high enantiomeric purity and 3,3-disubstituted-2,3-dihydrobenzofurans. Coupling with phenylacetylene in the presence of a Pd complex and CuI gives 2-phenylbenzofuran in good yield.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to light. Incompatible with oxidizing agents.
Specifications
| 2-Iodophenol | |
| 39°C to 41°C | |
| 186°C to 187°C (160 mmHg) | |
| C6H5IO | |
| 5 g | |
| Light sensitive | |
| o-iodophenol, phenol, 2-iodo, phenol, o-iodo, o-jodfenol, 2-jodfenol, phenol, iodo, o-jodfenol czech, 2-jodfenol czech, o-jodphenol, 2-iodanylphenol | |
| KQDJTBPASNJQFQ-UHFFFAOYSA-N | |
| 2-iodophenol | |
| 10784 | |
| 220.01 |
| 533-58-4 | |
| 1.947 | |
| >110°C (230°F) | |
| MFCD00013963 | |
| 1855300 | |
| 14,5035 | |
| Soluble in water. | |
| OC1=CC=CC=C1I | |
| 220.01 | |
| CHEBI:16706 | |
| 98% |
Chemical Identifiers
| 533-58-4 | |
| 220.01 | |
| KQDJTBPASNJQFQ-UHFFFAOYSA-N | |
| 10784 | |
| 2-iodophenol |
| C6H5IO | |
| MFCD00013963 | |
| o-iodophenol, phenol, 2-iodo, phenol, o-iodo, o-jodfenol, 2-jodfenol, phenol, iodo, o-jodfenol czech, 2-jodfenol czech, o-jodphenol, 2-iodanylphenol | |
| CHEBI:16706 | |
| OC1=CC=CC=C1I |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 208-569-9
RTECSNumber : SL5500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only