missing translation for 'onlineSavingsMsg'
Learn More

2-Iodophenol, 98%

Catalog No. p-149252
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC412480050 5 g Glass bottle
AC412480250 25 g Glass bottle
AC412481000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC412480050 Supplier Thermo Scientific Chemicals Supplier No. 412480050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 533-58-4 | C6H5IO | 220.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 533-58-4
Molecular Formula C6H5IO
Molecular Weight (g/mol) 220.01
MDL Number MFCD00013963
InChI Key KQDJTBPASNJQFQ-UHFFFAOYSA-N
Synonym o-iodophenol, phenol, 2-iodo, phenol, o-iodo, o-jodfenol, 2-jodfenol, phenol, iodo, o-jodfenol czech, 2-jodfenol czech, o-jodphenol, 2-iodanylphenol
PubChem CID 10784
ChEBI CHEBI:16706
IUPAC Name 2-iodophenol
SMILES OC1=CC=CC=C1I

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 37.0°C to 43.0°C
Color Brown to Cream or White
Boiling Point 186.0°C to 187.0°C (160.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula IC6H4OH
Quantity 5 g
Merck Index 15,5078
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol and ether
Formula Weight 220.01
Percent Purity 98%
Physical Form Crystalline Powder or Crystalline Low Melting Mass
Chemical Name or Material 2-Iodophenol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-569-9
RTECSNumber SL5500000
TSCA TSCA

RUO – Research Use Only