Learn More
2-Iodobiphenyl, 98%
CAS: 2113-51-1 | C12H9I | 280.108 g/mol
Supplier: Thermo Scientific Chemicals L0699006
Description
2-Iodobiphenyl is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Iodobiphenyl is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents.
Specifications
| 2-Iodobiphenyl | |
| 1.59 | |
| >110°C (230°F) | |
| 1.661 | |
| 5 g | |
| Light sensitive | |
| Insoluble in water. | |
| C1=CC=C(C=C1)C2=CC=CC=C2I | |
| 280.108 | |
| 280.11 |
| 2113-51-1 | |
| 300°C | |
| C12H9I | |
| MFCD00039396 | |
| 2206820 | |
| 2-iodobiphenyl, 2-iodo-1,1'-biphenyl, o-iodobiphenyl, 1,1'-biphenyl, 2-iodo, o-phenyliodobenzene, 2-iodo-biphenyl, biphenyl, 2-iodo, 2-iodo-1-phenylbenzene, 1-iodo-2-phenyl-benzene, 1,1'-biphenyl, iodo | |
| QFUYDAGNUJWBSM-UHFFFAOYSA-N | |
| 1-iodo-2-phenylbenzene | |
| 75025 | |
| 98% |
Chemical Identifiers
| 2113-51-1 | |
| 280.108 | |
| QFUYDAGNUJWBSM-UHFFFAOYSA-N | |
| 75025 | |
| C1=CC=C(C=C1)C2=CC=CC=C2I |
| C12H9I | |
| MFCD00039396 | |
| 2-iodobiphenyl, 2-iodo-1,1'-biphenyl, o-iodobiphenyl, 1,1'-biphenyl, 2-iodo, o-phenyliodobenzene, 2-iodo-biphenyl, biphenyl, 2-iodo, 2-iodo-1-phenylbenzene, 1-iodo-2-phenyl-benzene, 1,1'-biphenyl, iodo | |
| 1-iodo-2-phenylbenzene |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 218-303-3
RTECSNumber : DV5384800
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only