missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodobiphenyl, 98%
Description
2-Iodobiphenyl is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Iodobiphenyl is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents.
2-Iodobiphenyl is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 1.59 |
| Boiling Point | 300°C |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.661 |
| Quantity | 1 g |
| Beilstein | 2206820 |
| Sensitivity | Light sensitive |
| Solubility Information | Insoluble in water. |
| Formula Weight | 280.11 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 218-303-3 |
| RTECSNumber | DV5384800 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only