missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodobenzyl Alcohol 98.0+%, TCI America™
$109.74 - $329.20
Chemical Identifiers
| CAS | 5159-41-1 |
|---|---|
| Molecular Formula | C7H7IO |
| Molecular Weight (g/mol) | 234.036 |
| MDL Number | MFCD00004610 |
| InChI Key | WZCXOBMFBKSSFA-UHFFFAOYSA-N |
| Synonym | 2-iodobenzyl alcohol, 2-iodophenyl methanol, o-iodobenzyl alcohol, 2-iodobenzylalcohol, benzenemethanol, 2-iodo, 2-iodobenzylic alcohol, 2-jod-benzylalkohol, o-iodo benzylalcohol, 2-iodo-benzyl alcohol |
| PubChem CID | 107629 |
| IUPAC Name | (2-iodophenyl)methanol |
| SMILES | C1=CC=C(C(=C1)CO)I |
Chemical Identifiers
| 5159-41-1 | |
| 234.036 | |
| WZCXOBMFBKSSFA-UHFFFAOYSA-N | |
| 107629 | |
| C1=CC=C(C(=C1)CO)I |
| C7H7IO | |
| MFCD00004610 | |
| 2-iodobenzyl alcohol, 2-iodophenyl methanol, o-iodobenzyl alcohol, 2-iodobenzylalcohol, benzenemethanol, 2-iodo, 2-iodobenzylic alcohol, 2-jod-benzylalkohol, o-iodo benzylalcohol, 2-iodo-benzyl alcohol | |
| (2-iodophenyl)methanol |
Specifications
| 5159-41-1 | |
| White | |
| C7H7IO | |
| 5 g | |
| WZCXOBMFBKSSFA-UHFFFAOYSA-N | |
| (2-iodophenyl)methanol | |
| 107629 | |
| ≥98.0% (GC) | |
| 2-Iodobenzyl Alcohol |
| 92°C | |
| 145°C | |
| MFCD00004610 | |
| 2-iodobenzyl alcohol, 2-iodophenyl methanol, o-iodobenzyl alcohol, 2-iodobenzylalcohol, benzenemethanol, 2-iodo, 2-iodobenzylic alcohol, 2-jod-benzylalkohol, o-iodo benzylalcohol, 2-iodo-benzyl alcohol | |
| C1=CC=C(C(=C1)CO)I | |
| 234.036 | |
| 234.04 | |
| Crystalline Powder |
Safety and Handling
TSCA : No