missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzyl alcohol, 98+%

Catalog No. AAL1512606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1512606 5 g
AAL1512614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1512606 Supplier Thermo Scientific Chemicals Supplier No. L1512606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5159-41-1 | C7H7IO | 234.036 g/mol

2-Iodobenzyl alcohol was used in the synthesis of substituted seven-membered lactones, 2-[(E)-(1?-iodo-2?-propenyl)]benzyl alcohol2 and 2,3-diphenyl-1-indenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodobenzyl alcohol was used in the synthesis of substituted seven-membered lactones, 2-[(E)-(1′-iodo-2′-propenyl)]benzyl alcohol2 and 2,3-diphenyl-1-indenone.

Solubility
Insoluble in water.

Notes
Light Sensitive, store in dark. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 5159-41-1
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.036
MDL Number MFCD00004610
InChI Key WZCXOBMFBKSSFA-UHFFFAOYSA-N
Synonym 2-iodobenzyl alcohol, 2-iodophenyl methanol, o-iodobenzyl alcohol, 2-iodobenzylalcohol, benzenemethanol, 2-iodo, 2-iodobenzylic alcohol, 2-jod-benzylalkohol, o-iodo benzylalcohol, 2-iodo-benzyl alcohol
PubChem CID 107629
IUPAC Name (2-iodophenyl)methanol
SMILES C1=CC=C(C(=C1)CO)I

Specifications

Melting Point 89°C to 92°C
Quantity 5 g
Beilstein 2079487
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 234.04
Percent Purity 99%
Chemical Name or Material 2-Iodobenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-933-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only