missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodobenzoyl Chloride 97.0+%, TCI America™
$130.92 - $261.82
Chemical Identifiers
| CAS | 609-67-6 |
|---|---|
| Molecular Formula | C7H4ClIO |
| Molecular Weight (g/mol) | 266.462 |
| MDL Number | MFCD00001040 |
| InChI Key | MVIVDSWUOGNODP-UHFFFAOYSA-N |
| PubChem CID | 69112 |
| IUPAC Name | 2-iodobenzoyl chloride |
| SMILES | C1=CC=C(C(=C1)C(=O)Cl)I |
Chemical Identifiers
| 609-67-6 | |
| 266.462 | |
| MVIVDSWUOGNODP-UHFFFAOYSA-N | |
| 2-iodobenzoyl chloride |
| C7H4ClIO | |
| MFCD00001040 | |
| 69112 | |
| C1=CC=C(C(=C1)C(=O)Cl)I |
Specifications
| 609-67-6 | |
| C7H4ClIO | |
| 10 g | |
| o-iodobenzoyl chloride, benzoyl chloride, 2-iodo, iodobenzoyl chloride, o-iodobenzoylchloride, 2-iodobezoyl chloride, o-iodo-benzoylchloride, 2-iodobenzoic chloride, 2-iodo-benzoyl chloride, acmc-1atjg, benzoyl chloride,2-iodo | |
| C1=CC=C(C(=C1)C(=O)Cl)I | |
| 266.462 | |
| 266.46 | |
| Crystalline Lumps |
| 106°C | |
| MFCD00001040 | |
| 3261 | |
| MVIVDSWUOGNODP-UHFFFAOYSA-N | |
| 2-iodobenzoyl chloride | |
| 69112 | |
| ≥97.0% (T) | |
| 2-Iodobenzoyl Chloride |
Safety and Handling
TSCA : Yes