accessibility menu, dialog, popup

UserName

ClOI

CAS RN 609-67-6

IUPAC Name: 2-iodobenzoyl chloride
Synonyms: 2-iodobenzoyl chloride
Molecular Weight (g/mol): 266.46
Molecular Formula: C7H4ClIO
InChi Key: MVIVDSWUOGNODP-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=CC=C1I
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)