missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzoyl Chloride 97.0+%, TCI America™

Catalog No. I040210G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I040210G 10 g
I040225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I040210G Supplier TCI America Supplier No. I040210G
Only null left

Chemical Identifiers

CAS 609-67-6
Molecular Formula C7H4ClIO
Molecular Weight (g/mol) 266.462
MDL Number MFCD00001040
InChI Key MVIVDSWUOGNODP-UHFFFAOYSA-N
PubChem CID 69112
IUPAC Name 2-iodobenzoyl chloride
SMILES C1=CC=C(C(=C1)C(=O)Cl)I

Specifications

Boiling Point 106°C
Quantity 10 g
UN Number 3261
Synonym o-iodobenzoyl chloride, benzoyl chloride, 2-iodo, iodobenzoyl chloride, o-iodobenzoylchloride, 2-iodobezoyl chloride, o-iodo-benzoylchloride, 2-iodobenzoic chloride, 2-iodo-benzoyl chloride, acmc-1atjg, benzoyl chloride,2-iodo
Formula Weight 266.46
Percent Purity ≥97.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Iodobenzoyl Chloride