missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzoyl chloride, 98%

CAS: 609-67-6 | C7H4ClIO | 266.462 g/mol

$139.47 - $467.81

Chemical Identifiers

CAS 609-67-6
Molecular Formula C7H4ClIO
Molecular Weight (g/mol) 266.462
MDL Number MFCD00001040
InChI Key MVIVDSWUOGNODP-UHFFFAOYSA-N
PubChem CID 69112
IUPAC Name 2-iodobenzoyl chloride
SMILES C1=CC=C(C(=C1)C(=O)Cl)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1355809
View Documents
Thermo Scientific Chemicals
A1355809
10 g N/A
Only null left
 
AAA1355818
View Documents
Thermo Scientific Chemicals
A1355818
50 g N/A
Only null left
 
Description

Description

2-Iodobenzoyl chloride has been used in palladium-catalyzed synthesis of carbomethoxy functional group-induced isoindolin-1-ones, preparation of starting materials required for the synthesis of novel cyclic derivatives of pentavalent iodine, benziodazole oxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodobenzoyl chloride has been used in palladium-catalyzed synthesis of carbomethoxy functional group-induced isoindolin-1-ones, preparation of starting materials required for the synthesis of novel cyclic derivatives of pentavalent iodine, benziodazole oxides.

Solubility
Reacts with water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

609-67-6
266.462
MVIVDSWUOGNODP-UHFFFAOYSA-N
2-iodobenzoyl chloride
C7H4ClIO
MFCD00001040
69112
C1=CC=C(C(=C1)C(=O)Cl)I

Specifications

609-67-6
105°C to 106°C (1 mmHg)
C7H4ClIO
10 g
2042672
o-iodobenzoyl chloride, benzoyl chloride, 2-iodo, iodobenzoyl chloride, o-iodobenzoylchloride, 2-iodobezoyl chloride, o-iodo-benzoylchloride, 2-iodobenzoic chloride, 2-iodo-benzoyl chloride, acmc-1atjg, benzoyl chloride,2-iodo
MVIVDSWUOGNODP-UHFFFAOYSA-N
2-iodobenzoyl chloride
69112
98%
27°C to 32°C
>110°C (230°F)
MFCD00001040
UN3261
Moisture sensitive
Reacts with water.
C1=CC=C(C(=C1)C(=O)Cl)I
266.462
266.47
2-Iodobenzoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 210-196-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only