missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzoyl chloride, 98%

Catalog No. AAA1355809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1355809 10 g
AAA1355818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1355809 Supplier Thermo Scientific Chemicals Supplier No. A1355809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 609-67-6 | C7H4ClIO | 266.462 g/mol

2-Iodobenzoyl chloride has been used in palladium-catalyzed synthesis of carbomethoxy functional group-induced isoindolin-1-ones, preparation of starting materials required for the synthesis of novel cyclic derivatives of pentavalent iodine, benziodazole oxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodobenzoyl chloride has been used in palladium-catalyzed synthesis of carbomethoxy functional group-induced isoindolin-1-ones, preparation of starting materials required for the synthesis of novel cyclic derivatives of pentavalent iodine, benziodazole oxides.

Solubility
Reacts with water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 609-67-6
Molecular Formula C7H4ClIO
Molecular Weight (g/mol) 266.462
MDL Number MFCD00001040
InChI Key MVIVDSWUOGNODP-UHFFFAOYSA-N
PubChem CID 69112
IUPAC Name 2-iodobenzoyl chloride
SMILES C1=CC=C(C(=C1)C(=O)Cl)I

Specifications

Melting Point 27°C to 32°C
Boiling Point 105°C to 106°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
UN Number UN3261
Beilstein 2042672
Sensitivity Moisture sensitive
Synonym o-iodobenzoyl chloride, benzoyl chloride, 2-iodo, iodobenzoyl chloride, o-iodobenzoylchloride, 2-iodobezoyl chloride, o-iodo-benzoylchloride, 2-iodobenzoic chloride, 2-iodo-benzoyl chloride, acmc-1atjg, benzoyl chloride,2-iodo
Solubility Information Reacts with water.
Formula Weight 266.47
Percent Purity 98%
Chemical Name or Material 2-Iodobenzoyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 210-196-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only