missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodobenzoic acid, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 161.0°C to 163.0°C |
| Color | Brown-Orange to Yellow |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (HPLC) |
| Packaging | Plastic bottle |
| Linear Formula | IC6H4CO2H |
| Quantity | 25 g |
| Beilstein | 09,363 |
| Fieser | 01,506 |
| Merck Index | 15,5074 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes serious eye damage. Causes skin irritation. May cause respiratory irritation. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for |
| EINECSNumber | 201-850-7 |
RUO – Research Use Only