missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzoic acid, 98+%

Catalog No. AAA1056314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1056314 25 g
AAA1056322 100 g
AAA1056336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1056314 Supplier Thermo Scientific Chemicals Supplier No. A1056314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 88-67-5 | C7H5IO2 | 248.019 g/mol

Reagent for detection of sulfhydryl groups in proteins2-Iodobenzoic acid acts as a precursor in the preparation of 2-Iodoxybenzoic acid (IBX) and Dess-Martin periodinane, which finds application as an oxidizing agent in synthetic chemistry. It is also used in Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for detection of sulfhydryl groups in proteins2-Iodobenzoic acid acts as a precursor in the preparation of 2-Iodoxybenzoic acid (IBX) and Dess-Martin periodinane, which finds application as an oxidizing agent in synthetic chemistry. It is also used in Suzuki reaction.

Solubility
Soluble in dimethyl sulfoxide and methanol

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 88-67-5
Molecular Formula C7H5IO2
Molecular Weight (g/mol) 248.019
MDL Number MFCD00002419
InChI Key CJNZAXGUTKBIHP-UHFFFAOYSA-N
Synonym o-iodobenzoic acid, benzoic acid, 2-iodo, iodobenzoic acid, benzoic acid, o-iodo, usaf ek-572, 2-iodo benzoic acid, 2-iodo-benzoic acid, benzoic acid, iodo, kyselina o-jodbenzoova, o-jod-benzoesaeure
PubChem CID 6941
ChEBI CHEBI:287979
IUPAC Name 2-iodobenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)I

Specifications

Melting Point 160°C to 164°C
Density 2.25
Quantity 25 g
Beilstein 1861406
Sensitivity Light sensitive
Merck Index 14,5030
Solubility Information Soluble in dimethyl sulfoxide and methanol
Formula Weight 248.02
Percent Purity ≥98%
Chemical Name or Material 2-Iodobenzoic acid
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 201-850-7
RTECSNumber DH2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only