missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzaldehyde 98.0+%, TCI America™

Catalog No. p-7131513
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06101G 1 g
I06105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I06101G Supplier TCI America Supplier No. I06101G
Only null left

Chemical Identifiers

CAS 26260-02-6
Molecular Formula C7H5IO
Molecular Weight (g/mol) 232.02
MDL Number MFCD00039570
InChI Key WWKKTHALZAYYAI-UHFFFAOYSA-N
Synonym benzaldehyde, 2-iodo, o-iodobenzaldehyde, iodobenzaldehyde, benzaldehyde,iodo, 2-iodobenzenealdehyde, pubchem3914, acmc-209upi, 2-iodobenzaldehyde, ksc203m8l
PubChem CID 643439
IUPAC Name 2-iodobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)I

Specifications

Melting Point 39°C
Color White-Yellow
Boiling Point 102°C
Quantity 1 g
Formula Weight 232.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodobenzaldehyde
TSCA No
Recommended Storage Refrigerator