missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoaniline, 98%

CAS: 615-43-0 | C6H6IN | 219.03 g/mol

$91.67 - $91.67

Chemical Identifiers

CAS 615-43-0
Molecular Formula C6H6IN
Molecular Weight (g/mol) 219.03
MDL Number MFCD00007680
InChI Key UBPDKIDWEADHPP-UHFFFAOYSA-N
Synonym o-iodoaniline, benzenamine, 2-iodo, 2-iodo aniline, 2-iodobenzenamine, o-aminoiodobenzene, aniline, o-iodo, aniline, 2-iodo, benzenamine, iodo, 2-iodo-phenylamine, 2-iodophenylamine
PubChem CID 11995
IUPAC Name 2-iodoaniline
SMILES NC1=CC=CC=C1I
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC122300050
SDS View Documents
Thermo Scientific Chemicals
122300050
5 g Glass bottle
Each for $91.67
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

615-43-0
219.03
UBPDKIDWEADHPP-UHFFFAOYSA-N
11995
NC1=CC=CC=C1I
C6H6IN
MFCD00007680
o-iodoaniline, benzenamine, 2-iodo, 2-iodo aniline, 2-iodobenzenamine, o-aminoiodobenzene, aniline, o-iodo, aniline, 2-iodo, benzenamine, iodo, 2-iodo-phenylamine, 2-iodophenylamine
2-iodoaniline

Specifications

615-43-0
100.0
Brown to Yellow
Authentic
Glass bottle
MFCD00007680
12, 669
Solubility in water: slightly soluble
NC1=CC=CC=C1I
219.03
219.02
Crystalline Needles
97.5
53°C to 58°C
>110°C
98%
C6H6IN
5 g
o-iodoaniline, benzenamine, 2-iodo, 2-iodo aniline, 2-iodobenzenamine, o-aminoiodobenzene, aniline, o-iodo, aniline, 2-iodo, benzenamine, iodo, 2-iodo-phenylamine, 2-iodophenylamine
UBPDKIDWEADHPP-UHFFFAOYSA-N
2-iodoaniline
11995
98%
2-Iodoaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 210-426-

SDS
Documents

Documents

RUO – Research Use Only