missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoaniline, 98+%

Catalog No. p-7020935
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1305906 5 g
AAA1305914 25 g
AAA1305918 50 g
AAA1305922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1305906 Supplier Thermo Scientific Chemicals Supplier No. A1305906

CAS: 615-43-0 | C6H6IN | 219.03 g/mol

2-Iodoaniline is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodoaniline is used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
It is sensitive to light. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 615-43-0
Molecular Formula C6H6IN
Molecular Weight (g/mol) 219.03
MDL Number MFCD00007680
InChI Key UBPDKIDWEADHPP-UHFFFAOYSA-N
Synonym o-iodoaniline, benzenamine, 2-iodo, 2-iodo aniline, 2-iodobenzenamine, o-aminoiodobenzene, aniline, o-iodo, aniline, 2-iodo, benzenamine, iodo, 2-iodo-phenylamine, 2-iodophenylamine
PubChem CID 11995
IUPAC Name 2-iodoaniline
SMILES NC1=CC=CC=C1I

Specifications

Melting Point 53°C to 57°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2204899
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 219.03
Percent Purity ≥98%
Chemical Name or Material 2-Iodoaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-426-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only