missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-m-xylene 98.0+%, TCI America™

Catalog No. p-7131579
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07525G 5 g
I075225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07525G Supplier TCI America Supplier No. I07525G
Only null left

Chemical Identifiers

CAS 608-28-6
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.064
MDL Number MFCD00019012
InChI Key QTUGGVBKWIYQSS-UHFFFAOYSA-N
Synonym 2-iodo-m-xylene, 2,6-dimethyliodobenzene, m-xylene, 2-iodo, benzene, 2-iodo-1,3-dimethyl, 1,3-dimethyl-2-iodobenzene, 2-iodo-1,3-dimethyl-benzene, 2,6-dimethyl-1-iodobenzene, 2-iodo-o-xylene, pubchem3867, acmc-1axwc
PubChem CID 69097
IUPAC Name 2-iodo-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)I

Specifications

Melting Point 11°C
Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 230°C
Formula Weight 232.06
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Iodo-m-xylene