missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-m-xylene, 98%

Catalog No. p-7021201
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1343906 5 g
AAA1343914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1343906 Supplier Thermo Scientific Chemicals Supplier No. A1343906
Only null left

CAS: 608-28-6 | C8H9I | 232.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 608-28-6
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.064
MDL Number MFCD00019012
InChI Key QTUGGVBKWIYQSS-UHFFFAOYSA-N
Synonym 2-iodo-m-xylene, 2,6-dimethyliodobenzene, m-xylene, 2-iodo, benzene, 2-iodo-1,3-dimethyl, 1,3-dimethyl-2-iodobenzene, 2-iodo-1,3-dimethyl-benzene, 2,6-dimethyl-1-iodobenzene, 2-iodo-o-xylene, pubchem3867, acmc-1axwc
PubChem CID 69097
IUPAC Name 2-iodo-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)I

Specifications

Density 1.62
Quantity 5 g
Refractive Index 1.6025
Beilstein 1857414
Boiling Point 230°C to 231°C
Formula Weight 232.07
Percent Purity 98%
Chemical Name or Material 2-Iodo-m-xylene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only