missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-4-nitrotoluene, 97%

Catalog No. AAB2149114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2149114 25 g
AAB2149106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2149114 Supplier Thermo Scientific Chemicals Supplier No. B2149114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7745-92-8 | C7H6INO2 | 263.03 g/mol

2-Iodo-4-nitrotoluene is used as a pharmaceutical intermediate, chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodo-4-nitrotoluene is used as a pharmaceutical intermediate, chemical synthesis intermediate.

Solubility
Soluble in toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Light sensitive.

Chemical Identifiers

CAS 7745-92-8
Molecular Formula C7H6INO2
Molecular Weight (g/mol) 263.03
MDL Number MFCD00024329
InChI Key BUQSRXQJUZTIEW-UHFFFAOYSA-N
Synonym 2-iodo-4-nitrotoluene, benzene, 2-iodo-1-methyl-4-nitro, 2-iodo-1-methyl-4-nitro-benzene, pubchem3957, acmc-209pa2, toluene, 2-iodo-4-nitro, timtec-bb sbb005821
PubChem CID 82188
IUPAC Name 2-iodo-1-methyl-4-nitrobenzene
SMILES CC1=CC=C(C=C1I)[N+]([O-])=O

Specifications

Quantity 25 g
Sensitivity Light sensitive
Solubility Information Soluble in toluene.
Formula Weight 263.03
Percent Purity 97%
Chemical Name or Material 2-Iodo-4-nitrotoluene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-808-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only