missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-4-nitroaniline 98.0+%, TCI America™

Catalog No. p-7131640
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08781G 1 g
I08785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08781G Supplier TCI America Supplier No. I08781G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6293-83-0
Molecular Formula C6H5IN2O2
Molecular Weight (g/mol) 264.02
MDL Number MFCD00955758
InChI Key LOLSEMNGXKAZBZ-UHFFFAOYSA-N
PubChem CID 222629
IUPAC Name 2-iodo-4-nitroaniline
SMILES NC1=CC=C(C=C1I)[N+]([O-])=O

Specifications

Melting Point 107°C
Color Yellow
Quantity 1 g
Formula Weight 264.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodo-4-nitroaniline