accessibility menu, dialog, popup

UserName

NH2INOO

CAS RN 6293-83-0

IUPAC Name: 2-iodo-4-nitroaniline
Synonyms: 2-iodo-4-nitroaniline
Molecular Weight (g/mol): 264.02
Molecular Formula: C6H5IN2O2
InChi Key: LOLSEMNGXKAZBZ-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1I)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results