missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodo-4,6-dimethylaniline, 98%, Thermo Scientific Chemicals
Description
It is employed in the synthesis of benzothiazoles through copper-catalyzed one-pot three-component reactions with use of sodium hydrosulfide as a sulfur surrogate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in the synthesis of benzothiazoles through copper-catalyzed one-pot three-component reactions with use of sodium hydrosulfide as a sulfur surrogate.
Solubility
Insoluble in water.
Notes
Light sensitive. Store away from light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
It is employed in the synthesis of benzothiazoles through copper-catalyzed one-pot three-component reactions with use of sodium hydrosulfide as a sulfur surrogate.
Solubility
Insoluble in water.
Notes
Light sensitive. Store away from light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
Specifications
Specifications
| Melting Point | 64°C to 65°C |
| Quantity | 1 g |
| UN Number | UN2811 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 247.08 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Iodo-4,6-dimethylaniline |
Safety and Handling
| Hazard Category | H302+H332-H311-H315-H319 |
| Hazard Statement | GHS H Statement H311-H302-H332-H315-H319 Toxic in contact with skin. Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only