CAS RN 4102-54-9
IUPAC Name:
2-iodo-4,6-dimethylaniline
Synonyms:
2-iodo-4,6-dimethylaniline
Molecular Weight (g/mol):
247.08
Molecular Formula:
C8H10IN
InChi Key:
ODPOIEACUVQCBZ-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=C(N)C(I)=C1
1
–
1
of
1
results
1
Promotions Available
2-Iodo-4,6-dimethylaniline, 98%, Thermo Scientific Chemicals
2-Iodo-4,6-dimethylaniline, 98%, Thermo Scientific Chemicals
CAS: 4102-54-9 Molecular Formula: C8H10IN Molecular Weight (g/mol): 247.079 MDL Number: MFCD07779005 InChI Key: ODPOIEACUVQCBZ-UHFFFAOYSA-N Synonym: 2,4-dimethyl-6-iodoaniline,2-iodo-4,6-di-methylaniline,benzenamine,2-iodo-4,6-dimethyl PubChem CID: 14040290 IUPAC Name: 2-iodo-4,6-dimethylaniline SMILES: CC1=CC(=C(C(=C1)I)N)C