missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-4,6-dimethylaniline, 98%, Thermo Scientific Chemicals

Catalog No. AAH5187503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5187503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5187503 Supplier Thermo Scientific Chemicals Supplier No. H5187503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

It is employed in the synthesis of benzothiazoles through copper-catalyzed one-pot three-component reactions with use of sodium hydrosulfide as a sulfur surrogate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of benzothiazoles through copper-catalyzed one-pot three-component reactions with use of sodium hydrosulfide as a sulfur surrogate.

Solubility
Insoluble in water.

Notes
Light sensitive. Store away from light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.

Chemical Identifiers

CAS 4102-54-9
Molecular Formula C8H10IN
Molecular Weight (g/mol) 247.079
MDL Number MFCD07779005
InChI Key ODPOIEACUVQCBZ-UHFFFAOYSA-N
Synonym 2,4-dimethyl-6-iodoaniline, 2-iodo-4,6-di-methylaniline, benzenamine,2-iodo-4,6-dimethyl
PubChem CID 14040290
IUPAC Name 2-iodo-4,6-dimethylaniline
SMILES CC1=CC(=C(C(=C1)I)N)C

Specifications

Melting Point 64°C to 65°C
Quantity 1 g
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 247.08
Percent Purity 98%
Chemical Name or Material 2-Iodo-4,6-dimethylaniline
Hazard Category H302+H332-H311-H315-H319
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only