missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodo-2-methylpropionitrile 96.0+%, TCI America™
Supplier: TCI America I09541G
Specifications
| 2-Iodo-2-methylpropionitrile | |
| 62°C | |
| MFCD28384148 | |
| 3276 | |
| CC(C)(I)C#N | |
| 195.00 | |
| 195.00 | |
| Liquid |
| 19481-79-9 | |
| C4H6IN | |
| 1 g | |
| DBESYUOJKJZGLV-UHFFFAOYSA-N | |
| 2-iodo-2-methylpropanenitrile | |
| 549274 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 19481-79-9 | |
| 195.00 | |
| DBESYUOJKJZGLV-UHFFFAOYSA-N | |
| 2-iodo-2-methylpropanenitrile |
| C4H6IN | |
| MFCD28384148 | |
| 549274 | |
| CC(C)(I)C#N |
Safety and Handling
TSCA : No
Recommended Storage : Freezer