missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-2-methylpropionitrile 96.0+%, TCI America™

Catalog No. p-7131676
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09541G 1 g
I09545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09541G Supplier TCI America Supplier No. I09541G
Only null left

Chemical Identifiers

CAS 19481-79-9
Molecular Formula C4H6IN
Molecular Weight (g/mol) 195.00
MDL Number MFCD28384148
InChI Key DBESYUOJKJZGLV-UHFFFAOYSA-N
PubChem CID 549274
IUPAC Name 2-iodo-2-methylpropanenitrile
SMILES CC(C)(I)C#N

Specifications

Boiling Point 62°C
Quantity 1 g
UN Number 3276
Formula Weight 195.00
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Iodo-2-methylpropionitrile
TSCA No
Recommended Storage Freezer