accessibility menu, dialog, popup

UserName

CAS RN 19481-79-9

IUPAC Name: 2-iodo-2-methylpropanenitrile
Synonyms: 2-iodo-2-methylpropanenitrile
Molecular Weight (g/mol): 195
Molecular Formula: C4H6IN
InChi Key: DBESYUOJKJZGLV-UHFFFAOYSA-N
SMILES: CC(C)(I)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results