Learn More
2-Indanylboronic acid pinacol ester, 95%
CAS: 608534-44-7 | C15H21BO2 | 244.14 g/mol
Supplier: Thermo Scientific Chemicals L1953503
Description
Boronic acids react with alcohols, with loss of water, to form boronic esters. With simple alcohols, the products are very susceptible to hydrolysis, but with 1,2- and 1,3-diols, the resulting cyclic boronates (1,3,2- dioxaborolanes and 1,3,2-dioxaborinanes) are stable enough to be isolated.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBoronic acids react with alcohols, with loss of water, to form boronic esters. With simple alcohols, the products are very susceptible to hydrolysis, but with 1,2- and 1,3-diols, the resulting cyclic boronates (1,3,2- dioxaborolanes and 1,3,2-dioxaborinanes) are stable enough to be isolated.
Solubility
Difficult to mix in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications
| 2-Indanylboronic acid pinacol ester | |
| C15H21BO2 | |
| MFCD03788726 | |
| 2-indanylboronic acid pinacol ester, 2-2,3-dihydro-1h-inden-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-indanylboronicacidpinacolester, 2-indan-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| YRTKSYXHFWUPAZ-UHFFFAOYSA-N | |
| 2-(2,3-dihydro-1H-inden-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 24208876 | |
| 95% |
| 608534-44-7 | |
| 1.504 | |
| 1 g | |
| Difficult to mix in water. | |
| CC1(C)OB(OC1(C)C)C1CC2=CC=CC=C2C1 | |
| 244.14 | |
| 244.14 |
Chemical Identifiers
| 608534-44-7 | |
| 244.14 | |
| YRTKSYXHFWUPAZ-UHFFFAOYSA-N | |
| 24208876 | |
| CC1(C)OB(OC1(C)C)C1CC2=CC=CC=C2C1 |
| C15H21BO2 | |
| MFCD03788726 | |
| 2-indanylboronic acid pinacol ester, 2-2,3-dihydro-1h-inden-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-indanylboronicacidpinacolester, 2-indan-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 2-(2,3-dihydro-1H-inden-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only