accessibility menu, dialog, popup

UserName

CAS RN 608534-44-7

IUPAC Name: 2-(2,3-dihydro-1H-inden-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(2,3-dihydro-1H-inden-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 244.14
Molecular Formula: C15H21BO2
InChi Key: YRTKSYXHFWUPAZ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1CC2=CC=CC=C2C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)