missing translation for 'onlineSavingsMsg'
Learn More

2-Indanylboronic acid pinacol ester, 95%

Catalog No. AAL19535MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19535MD 250 mg
AAL1953503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19535MD Supplier Thermo Scientific Chemicals Supplier No. L19535MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 608534-44-7 | C15H21BO2 | 244.14 g/mol

Boronic acids react with alcohols, with loss of water, to form boronic esters. With simple alcohols, the products are very susceptible to hydrolysis, but with 1,2- and 1,3-diols, the resulting cyclic boronates (1,3,2- dioxaborolanes and 1,3,2-dioxaborinanes) are stable enough to be isolated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boronic acids react with alcohols, with loss of water, to form boronic esters. With simple alcohols, the products are very susceptible to hydrolysis, but with 1,2- and 1,3-diols, the resulting cyclic boronates (1,3,2- dioxaborolanes and 1,3,2-dioxaborinanes) are stable enough to be isolated.

Solubility
Difficult to mix in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 608534-44-7
Molecular Formula C15H21BO2
Molecular Weight (g/mol) 244.14
MDL Number MFCD03788726
InChI Key YRTKSYXHFWUPAZ-UHFFFAOYSA-N
Synonym 2-indanylboronic acid pinacol ester, 2-2,3-dihydro-1h-inden-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-indanylboronicacidpinacolester, 2-indan-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 24208876
IUPAC Name 2-(2,3-dihydro-1H-inden-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1CC2=CC=CC=C2C1

Specifications

Refractive Index 1.504
Quantity 250 mg
Solubility Information Difficult to mix in water.
Formula Weight 244.14
Percent Purity 95%
Chemical Name or Material 2-Indanylboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only