missing translation for 'onlineSavingsMsg'
Learn More

2-Indanylacetic acid, 99%

Catalog No. p-7045433
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1070003 1 g
AAL1070006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1070003 Supplier Thermo Scientific Chemicals Supplier No. L1070003
Only null left

CAS: 37868-26-1 | C11H12O2 | 176.22 g/mol

2-Indanylacetic acid is the starting material in the synthesis of pentacylic core of (+)-Salvileucalin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Indanylacetic acid is the starting material in the synthesis of pentacylic core of (+)-Salvileucalin.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 37868-26-1
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.22
MDL Number MFCD00082653
InChI Key TULDPXYHBFBRGW-UHFFFAOYSA-N
Synonym 2-2,3-dihydro-1h-inden-2-yl acetic acid, indan-2-yl-acetic acid, 1h-indene-2-acetic acid, 2,3-dihydro, 2,3-dihydro-1h-inden-2-ylacetic acid, 2,3-dihydro-1h-indene-2-acetic acid, 2-indaneacetic acid, indan-2-acetic acid, pubchem16311, indan-2-yl acetic acid
PubChem CID 5174955
IUPAC Name 2-(2,3-dihydro-1H-inden-2-yl)acetic acid
SMILES OC(=O)CC1CC2=CC=CC=C2C1

Specifications

Melting Point 91°C to 93°C
Quantity 1 g
Beilstein 2616409
Solubility Information Insoluble in water.
Formula Weight 176.22
Percent Purity 99%
Chemical Name or Material 2-Indanylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only