CAS RN 37868-26-1
IUPAC Name:
2-(2,3-dihydro-1H-inden-2-yl)acetic acid
Synonyms:
2,3-dihydro-1H-inden-2-ylacetic acid
Molecular Weight (g/mol):
176.22
Molecular Formula:
C11H12O2
InChi Key:
TULDPXYHBFBRGW-UHFFFAOYSA-N
SMILES:
OC(=O)CC1CC2=CC=CC=C2C1
1
–
1
of
1
results
1
Promotions Available
2-Indanylacetic acid, 99%
2-Indanylacetic acid, 99%
CAS: 37868-26-1 Molecular Formula: C11H12O2 Molecular Weight (g/mol): 176.22 MDL Number: MFCD00082653 InChI Key: TULDPXYHBFBRGW-UHFFFAOYSA-N Synonym: 2-2,3-dihydro-1h-inden-2-yl acetic acid,indan-2-yl-acetic acid,1h-indene-2-acetic acid, 2,3-dihydro,2,3-dihydro-1h-inden-2-ylacetic acid,2,3-dihydro-1h-indene-2-acetic acid,2-indaneacetic acid,indan-2-acetic acid,pubchem16311,indan-2-yl acetic acid PubChem CID: 5174955 IUPAC Name: 2-(2,3-dihydro-1H-inden-2-yl)acetic acid SMILES: OC(=O)CC1CC2=CC=CC=C2C1