missing translation for 'onlineSavingsMsg'
Learn More

2-Indancarboxylic Acid 98.0+%, TCI America™

Catalog No. I05321G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05321G 1 g
I05325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05321G Supplier TCI America Supplier No. I05321G

Chemical Identifiers

CAS 25177-85-9
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00085095
InChI Key XUDCMQBOWOLYCF-UHFFFAOYSA-N
Synonym 2-indancarboxylic acid, indane-2-carboxylic acid, indan-2-carboxylic acid, 2-indanecarboxylic acid, 1h-indene-2-carboxylic acid, 2,3-dihydro, chembl81558, 2-carboxyindane, 2-carboxy-indan, 2-carboxy-indane, 2-indancarboxylicacid
PubChem CID 575777
IUPAC Name 2,3-dihydro-1H-indene-2-carboxylic acid
SMILES OC(=O)C1CC2=CC=CC=C2C1

Specifications

Melting Point 128°C
Boiling Point 192°C
Quantity 1 g
Formula Weight 162.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Indancarboxylic Acid