accessibility menu, dialog, popup

UserName

CAS RN 25177-85-9

IUPAC Name: 2,3-dihydro-1H-indene-2-carboxylic acid
Synonyms: 2,3-dihydro-1H-indene-2-carboxylic acid
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: XUDCMQBOWOLYCF-UHFFFAOYSA-N
SMILES: OC(=O)C1CC2=CC=CC=C2C1
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)