missing translation for 'onlineSavingsMsg'
Learn More

2-Imidazolidone hemihydrate, 99+%

Catalog No. p-3778855
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC122045000 500 g
AC122040025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC122045000 Supplier Thermo Scientific Chemicals Supplier No. 122045000
Only null left

CAS: 121325-67-5 | C6H14N4O3 | 190.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121325-67-5 , 120-93-4
Molecular Formula C6H14N4O3
Molecular Weight (g/mol) 190.20
MDL Number MFCD02662355
InChI Key KPRJGGOOWATRNT-UHFFFAOYSA-N
Synonym imidazolidin-2-one hydrate, 2-imidazolidinone hemihydrate, 2-imidazolidone hemihydrate, bis imidazolidin-2-one hydrate, bis imidazolidinone hydrate, 2-imidazolidinone, hydrate 2:1, imidazolidin-2-one, imidazolidin-2-one, hydrate
PubChem CID 2723651
IUPAC Name imidazolidin-2-one;hydrate
SMILES O.O=C1NCCN1.O=C1NCCN1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 58.0°C
Color White to Yellow
Density 265g/mL
Boiling Point >100.0°C
Flash Point 265°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Quantity 500 g
Merck Index 14, 4914
Solubility Information Solubility in water: 950g/L (20°C). Other solubilities: 1800g/L water (40°C)
Formula Weight 95.1
Percent Purity 99+%
Physical Form Adhering Crystals or Crystalline Powder
Chemical Name or Material 2-Imidazolidone hemihydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Suspected of damaging fertility.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only