accessibility menu, dialog, popup

UserName

CAS RN 121325-67-5

IUPAC Name: bis(imidazolidin-2-one) hydrate
Synonyms: bis(imidazolidinone) hydrate
Molecular Weight (g/mol): 190.2
Molecular Formula: C6H14N4O3
InChi Key: KPRJGGOOWATRNT-UHFFFAOYSA-N
SMILES: O.O=C1NCCN1.O=C1NCCN1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)