accessibility menu, dialog, popup

UserName

CAS RN 120-93-4

IUPAC Name: imidazolidin-2-one
Synonyms: imidazolidinone|ethylene urea|ethyleneurea|N,N'-ethyleneurea|urea, N,N'-(1,2-ethanediyl)-|urea, 1,3-ethylene-|monoethyleneurea|2-oxomidazolidine|1,3-ethyleneurea|imidazolidinone (product)
Molecular Weight (g/mol): 86.09
Molecular Formula: C3H6N2O
InChi Key: YAMHXTCMCPHKLN-UHFFFAOYSA-N
SMILES: O=C1NCCN1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)