missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Hydroxymethyl)phenylboronic Acid Cyclic Monoester 97.0+%, TCI America™
Supplier: TCI America H12805G
Specifications
| 2-(Hydroxymethyl)phenylboronic Acid Cyclic Monoester | |
| 98°C | |
| C7H7BO2 | |
| 5 g | |
| XOQABDOICLHPIS-UHFFFAOYSA-N | |
| 1,3-dihydro-2,1-benzoxaborol-1-ol | |
| 403788 | |
| 133.94 |
| 5735-41-1 | |
| White | |
| MFCD01075677 | |
| 1-hydroxy-2,1-benzoxaborolane, benzo c 1,2 oxaborol-1 3h-ol, 2,1-benzoxaborol-1 3h-ol, 1,3-dihydro-1-hydroxy-2,1-benzoxaborole, 1,3-dihydro-2,1-benzoxaborol-1-ol, monoester, 2-hydroxymethyl phenylboronic acid cyclic monoester, 2-hydroxymethyl phenylboronic acid, dehydrate, 3h-2,1-benzoxaborol-1-ol, 2,1-benzoxaborole, 1,3-dihydro-1-hydroxy | |
| OB1OCC2=CC=CC=C12 | |
| 133.94 | |
| CHEBI:78238 | |
| Crystalline Powder |
Chemical Identifiers
| 5735-41-1 | |
| 133.94 | |
| XOQABDOICLHPIS-UHFFFAOYSA-N | |
| 403788 | |
| 1,3-dihydro-2,1-benzoxaborol-1-ol |
| C7H7BO2 | |
| MFCD01075677 | |
| 1-hydroxy-2,1-benzoxaborolane, benzo c 1,2 oxaborol-1 3h-ol, 2,1-benzoxaborol-1 3h-ol, 1,3-dihydro-1-hydroxy-2,1-benzoxaborole, 1,3-dihydro-2,1-benzoxaborol-1-ol, monoester, 2-hydroxymethyl phenylboronic acid cyclic monoester, 2-hydroxymethyl phenylboronic acid, dehydrate, 3h-2,1-benzoxaborol-1-ol, 2,1-benzoxaborole, 1,3-dihydro-1-hydroxy | |
| CHEBI:78238 | |
| OB1OCC2=CC=CC=C12 |
Safety and Handling
TSCA : No