missing translation for 'onlineSavingsMsg'
Learn More

2-(Hydroxymethyl)phenylboronic Acid Cyclic Monoester 97.0+%, TCI America™

Catalog No. p-7130957
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12801G 1 g
H12805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12801G Supplier TCI America Supplier No. H12801G
Only null left

Chemical Identifiers

CAS 5735-41-1
Molecular Formula C7H7BO2
Molecular Weight (g/mol) 133.94
MDL Number MFCD01075677
InChI Key XOQABDOICLHPIS-UHFFFAOYSA-N
Synonym 1-hydroxy-2,1-benzoxaborolane, benzo c 1,2 oxaborol-1 3h-ol, 2,1-benzoxaborol-1 3h-ol, 1,3-dihydro-1-hydroxy-2,1-benzoxaborole, 1,3-dihydro-2,1-benzoxaborol-1-ol, monoester, 2-hydroxymethyl phenylboronic acid cyclic monoester, 2-hydroxymethyl phenylboronic acid, dehydrate, 3h-2,1-benzoxaborol-1-ol, 2,1-benzoxaborole, 1,3-dihydro-1-hydroxy
PubChem CID 403788
ChEBI CHEBI:78238
IUPAC Name 1,3-dihydro-2,1-benzoxaborol-1-ol
SMILES OB1OCC2=CC=CC=C12

Specifications

Melting Point 98°C
Color White
Quantity 1 g
Formula Weight 133.94
Physical Form Crystalline Powder
Chemical Name or Material 2-(Hydroxymethyl)phenylboronic Acid Cyclic Monoester