accessibility menu, dialog, popup

UserName

CAS RN 5735-41-1

IUPAC Name: 1,3-dihydro-2,1-benzoxaborol-1-ol
Synonyms: 3H-2,1-benzoxaborol-1-ol
Molecular Weight (g/mol): 133.94
Molecular Formula: C7H7BO2
InChi Key: XOQABDOICLHPIS-UHFFFAOYSA-N
SMILES: OB1OCC2=CC=CC=C12
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results