missing translation for 'onlineSavingsMsg'
Learn More

(2-Hydroxyethyl)triphenylphosphonium bromide, 98+%

Catalog No. AAL0067922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0067922 100 g
AAL0067914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0067922 Supplier Thermo Scientific Chemicals Supplier No. L0067922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7237-34-5 | C20H20BrOP | 387.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7237-34-5
Molecular Formula C20H20BrOP
Molecular Weight (g/mol) 387.26
MDL Number MFCD00011901
InChI Key QZJOQNHOOVSESC-UHFFFAOYSA-M
Synonym 2-hydroxyethyl triphenylphosphonium bromide, 2-hydroxyethyl triphenylphosphanium bromide, acmc-20appe, 2-hydroxyethyltriphenylphosphonium bromide, 2-hydroxyethyl triphenyphosphonium bromide, 2-hydroxyethyl triphenyl phosphanium bromide, 2-hydroxy ethyl triphenylphosphonium bromide, phosphonium, 2-hydroxyethyl triphenyl-,bromide 1:1, phosphonium, 2-hydroxyethyl triphenyl-, bromide 1:1
PubChem CID 2733550
IUPAC Name 2-hydroxyethyl(triphenyl)phosphanium;bromide
SMILES [Br-].OCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 218°C to 220°C
Quantity 100 g
Beilstein 3922741
Sensitivity Hygroscopic
Formula Weight 387.26
Percent Purity ≥98%
Chemical Name or Material (2-Hydroxyethyl)triphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only